The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018827.2 | NP_061297.1 | 1 - 2 | 248 | 102 |
GenBank mRNA | AB040038.1 | - | 1 - 2 | 248 | 102 |