Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cbln1
- Entrez Gene ID
-
12404
- Gene Name
- cerebellin 1 precursor protein
- Gene Aliases
-
AI323299
- Chromosome Location
-
Chr.8: 87468853 - 87472592 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X61448.1 |
- |
1 - 2 |
262 |
86 |
View Full Product Details