Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crybb3
Entrez Gene ID
12962
Gene Name
crystallin, beta B3
Gene Aliases
AI852419
Chromosome Location
Chr.5: 113075839 - 113082233 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159650.1 NP_001153122.1 2 - 3 128 82
NM_021352.3 NP_067327.1 2 - 3 198 82
GenBank mRNA AJ272229.2 - 1 - 2 182 82
BC031442.1 - 1 - 2 189 82
BC058522.1 - 1 - 2 107 82
CJ046471.1 - 2 - 3 199 82

View Full Product Details