The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021541.3 | NP_067516.1 | 2 - 3 | 236 | 69 |
GenBank mRNA | AJ272227.1 | - | 2 - 3 | 228 | 69 |
AK021374.1 | - | 2 - 3 | 210 | 69 | |
AY160973.1 | - | 2 - 3 | 201 | 69 | |
BC052746.1 | - | 2 - 3 | 199 | 69 | |
BC056989.1 | - | 2 - 3 | 190 | 69 | |
CF740624.1 | - | 2 - 3 | 235 | 69 |