Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmfg
Entrez Gene ID
63986
Gene Name
glia maturation factor, gamma
Gene Aliases
0610039G16Rik, 2310057N07Rik, AI324845
Chromosome Location
Chr.7: 28437447 - 28448221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039192.1 NP_001034281.1 2 - 3 202 65
NM_022024.2 NP_071307.1 2 - 3 225 65
GenBank mRNA AB003588.1 - 2 - 3 159 65
AF297221.1 - 2 - 3 222 65
AK002834.1 - 2 - 3 226 65
AK009967.1 - 2 - 3 202 65
AK014214.1 - 2 - 3 226 65
BC011488.1 - 2 - 3 178 65

View Full Product Details