The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026412.3 | NP_080688.2 | 2 - 3 | 399 | 118 |
GenBank mRNA | AK006328.1 | - | 2 - 3 | 400 | 118 |
AK012148.1 | - | 2 - 3 | 199 | 118 | |
AK028590.1 | - | 2 - 3 | 200 | 118 | |
AK036374.1 | - | 2 - 3 | 188 | 118 | |
BC031709.1 | - | 1 - 2 | 107 | 118 |