The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026623.3 | NP_080899.1 | 6 - 7 | 810 | 90 |
GenBank mRNA | AK011688.2 | - | 6 - 7 | 800 | 90 |
AK012308.1 | - | 6 - 7 | 784 | 90 | |
AK019433.1 | - | 6 - 7 | 793 | 90 | |
AK020150.1 | - | 6 - 7 | 791 | 90 | |
AK146419.1 | - | 6 - 7 | 776 | 90 | |
AK147061.1 | - | - | 793 | 90 | |
AK160147.1 | - | 6 - 7 | 776 | 90 | |
AK213941.1 | - | 1 - 2 | 64 | 90 | |
BC008270.1 | - | 6 - 7 | 793 | 90 | |
BC090834.1 | - | - | 769 | 90 |