The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037846.3 | NP_001032935.2 | 13 - 14 | 1447 | 96 |
NM_001037847.2 | NP_001032936.2 | 13 - 14 | 1377 | 96 | |
NM_028082.2 | NP_082358.2 | 12 - 13 | 1324 | 96 | |
NR_024327.1 | 11 - 12 | 1257 | 96 | ||
GenBank mRNA | AK011231.1 | - | 11 - 12 | 1247 | 96 |
AK078121.1 | - | 13 - 14 | 1437 | 96 | |
AK149767.1 | - | 13 - 14 | 1421 | 96 | |
AK167834.1 | - | 13 - 14 | 1425 | 96 | |
BC043133.1 | - | 13 - 14 | 1348 | 96 | |
BC063105.1 | - | 14 - 15 | 1590 | 96 | |
BC065171.1 | - | 13 - 14 | 1377 | 96 | |
BC090624.1 | - | 11 - 12 | 1253 | 96 |