Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cwc22
Entrez Gene ID
80744
Gene Name
CWC22 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
AA684037, AI173004, AL022752, B230213M24, mKIAA1604
Chromosome Location
Chr.2: 77895653 - 77946356 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290740.1 NP_001277669.1 11 - 12 1521 78
NM_030560.5 NP_085037.2 12 - 13 1579 78
NM_172667.2 NP_766255.1 11 - 12 1518 78
GenBank mRNA AK045589.1 - 11 - 12 1350 78
AK077961.1 - 12 - 13 1407 78
AK129404.1 - - 1335 78
AK132001.1 - 11 - 12 1462 78
AK149528.1 - 11 - 12 1345 78
BC003273.1 - 5 - 6 506 78
BC003993.1 - 11 - 12 1331 78

View Full Product Details