Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
70314
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
2610011A08Rik, AI462746, Fra
Chromosome Location
Chr.7: 126428416 - 126445907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030566.2 NP_085043.2 5 - 6 1220 64
GenBank mRNA AK029188.1 - 3 - 4 64
AK089938.1 - 5 - 6 924 64
AK144836.1 - 5 - 6 906 64
AK157832.1 - 5 - 6 881 64
BC004088.1 - 4 - 5 966 64
BC015287.1 - 5 - 6 1228 64

View Full Product Details