Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr84
Entrez Gene ID
80910
Gene Name
G protein-coupled receptor 84
Gene Aliases
EX33
Chromosome Location
Chr.15: 103308233 - 103311242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030720.1 NP_109645.1 1 - 2 124 111
GenBank mRNA AF272948.1 - 1 - 2 124 111
BC023249.1 - 1 - 2 133 111

View Full Product Details