The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127338.1 | NP_001120810.1 | 18 - 19 | 1756 | 85 |
NM_138600.4 | NP_613066.2 | 16 - 17 | 1562 | 85 | |
GenBank mRNA | AK004991.1 | - | 18 - 19 | 1690 | 85 |
AK145873.1 | - | 16 - 17 | 1505 | 85 | |
AK160117.1 | - | 16 - 17 | 1489 | 85 | |
AK169195.1 | - | 16 - 17 | 1505 | 85 | |
BC012407.1 | - | 16 - 17 | 1524 | 85 |