Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Alg1
- Entrez Gene ID
-
208211
- Gene Name
- asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase)
- Gene Aliases
-
HMAT1, HMT1
- Chromosome Location
-
Chr.16: 5233621 - 5244907 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC011281.1 |
- |
1 - 2 |
218 |
85 |
View Full Product Details