The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145369.3 | NP_663344.2 | 1 - 2 | 176 | 75 |
GenBank mRNA | AK131905.1 | - | 1 - 2 | 124 | 75 |
AK131906.1 | - | 1 - 2 | 109 | 75 | |
AY669505.1 | - | 1 - 2 | 175 | 75 | |
BC019734.1 | - | 1 - 2 | 176 | 75 |