Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasgef1b
Entrez Gene ID
320292
Gene Name
RasGEF domain family, member 1B
Gene Aliases
4732452O09Rik, GPIgamma4, Gpig4
Chromosome Location
Chr.5: 99217420 - 99252927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145839.2 NP_665838.1 3 - 4 516 74
NM_181318.4 NP_851835.1 3 - 4 516 74
GenBank mRNA AK028756.1 - 3 - 4 408 74
AK090085.1 - 3 - 4 408 74
AK149812.1 - 3 - 4 435 74
AY129963.1 - 3 - 4 443 74
BC050858.1 - 3 - 4 449 74
BC052726.1 - 3 - 4 407 74

View Full Product Details