Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fntb
Entrez Gene ID
110606
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
2010013E13Rik, AA409500
Chromosome Location
Chr.12: 76837398 - 76921412 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145927.2 NP_666039.1 6 - 7 647 152
GenBank mRNA AK089911.1 - 6 - 7 677 152
BC031417.1 - 6 - 7 647 152

View Full Product Details