The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145934.2 | NP_666046.1 | 12 - 13 | 1242 | 82 |
NR_102313.1 | 11 - 12 | 1346 | 82 | ||
GenBank mRNA | AK050131.1 | - | 12 - 13 | 1222 | 82 |
AK050172.1 | - | 11 - 12 | 1314 | 82 | |
BC026642.1 | - | 12 - 13 | 1216 | 82 |