The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077398.2 | NP_001070866.1 | 4 - 5 | 785 | 121 |
NM_001286348.1 | NP_001273277.1 | 4 - 5 | 785 | 121 | |
NM_010698.4 | NP_034828.3 | 4 - 5 | 785 | 121 | |
GenBank mRNA | AB221652.1 | - | 4 - 5 | 532 | 121 |
AK146860.1 | - | 4 - 5 | 719 | 121 | |
AK147160.1 | - | 4 - 5 | 695 | 121 | |
BC079611.1 | - | 4 - 5 | 678 | 121 | |
U89487.1 | - | 4 - 5 | 674 | 121 | |
U89489.1 | - | 4 - 5 | 674 | 121 |