The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077398.2 | NP_001070866.1 | 6 - 7 | 991 | 78 |
NM_001286348.1 | NP_001273277.1 | 6 - 7 | 991 | 78 | |
NM_010698.4 | NP_034828.3 | 6 - 7 | 991 | 78 | |
GenBank mRNA | AK146860.1 | - | 6 - 7 | 925 | 78 |
AK147160.1 | - | 6 - 7 | 901 | 78 | |
BC079611.1 | - | 6 - 7 | 884 | 78 | |
U89487.1 | - | 6 - 7 | 880 | 78 | |
U89489.1 | - | 6 - 7 | 880 | 78 |