The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135688.1 | NP_001129160.1 | 3 - 4 | 389 | 65 |
NM_001135689.1 | NP_001129161.1 | 3 - 4 | 303 | 65 | |
NM_011837.3 | NP_035967.1 | 3 - 4 | 666 | 65 | |
GenBank mRNA | AF127091.1 | - | 3 - 4 | 299 | 65 |
AK021002.1 | - | 3 - 4 | 343 | 65 | |
AK034884.1 | - | - | 306 | 65 | |
BB627076.1 | - | 3 - 4 | 306 | 65 | |
BC028758.1 | - | 3 - 4 | 300 | 65 | |
BM943584.1 | - | 3 - 4 | 389 | 65 | |
BM948855.1 | - | 3 - 4 | 666 | 65 | |
CX238172.1 | - | 3 - 4 | 369 | 65 |