Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
208449
Gene Name
sphingomyelin synthase 1
Gene Aliases
9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
Chromosome Location
Chr.19: 32122727 - 32389705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168525.1 NP_001161997.1 9 - 10 2240 80
NM_001168526.1 NP_001161998.1 5 - 6 1464 80
NM_144792.4 NP_659041.3 9 - 10 1808 80
GenBank mRNA AK076554.1 - 9 - 10 1608 80
AK079254.1 - 5 - 6 1409 80
AK133967.1 - 5 - 6 1432 80
AK146782.1 - 9 - 10 1808 80
AK150272.1 - 5 - 6 1441 80
AK151170.1 - 9 - 10 1793 80
AK152312.1 - 5 - 6 1442 80
AK153406.1 - 9 - 10 1793 80
AK160539.1 - 5 - 6 1442 80
AY509044.1 - 9 - 10 1808 80
BC019443.1 - 5 - 6 1431 80
BC076618.1 - 9 - 10 2240 80
BC085298.1 - 6 - 7 1402 80

View Full Product Details