The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144879.2 | NP_659128.2 | 4 - 5 | 845 | 71 |
NR_027352.1 | 4 - 5 | 712 | 71 | ||
GenBank mRNA | AK045261.1 | - | 4 - 5 | 641 | 71 |
AK078596.1 | - | 4 - 5 | 714 | 71 | |
AK140989.1 | - | 4 - 5 | 839 | 71 | |
AK163193.1 | - | 4 - 5 | 621 | 71 | |
BC024141.1 | - | 4 - 5 | 615 | 71 |