Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zdhhc5
Entrez Gene ID
228136
Gene Name
zinc finger, DHHC domain containing 5
Gene Aliases
1110032A17Rik, AI451382, Zisp
Chromosome Location
Chr.2: 84687920 - 84715164 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144887.4 NP_659136.1 5 - 6 1791 109
GenBank mRNA AK028737.1 - 2 - 3 357 109
AK082513.1 - 5 - 6 1716 109
AK145086.1 - 5 - 6 1718 109
AK173251.1 - 5 - 6 1746 109
AY894891.1 - 5 - 6 1783 109
BC020051.1 - 4 - 5 761 109
BC065155.1 - 5 - 6 1661 109

View Full Product Details