Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35c2
Entrez Gene ID
228875
Gene Name
solute carrier family 35, member C2
Gene Aliases
C85957, CGI-15, D2Wsu58e, Ovcov1
Chromosome Location
Chr.2: 165276522 - 165287888 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045544.1 2 - 3 132 95
GenBank mRNA BC018327.1 - 2 - 3 121 95

View Full Product Details