Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnrnpul1
Entrez Gene ID
232989
Gene Name
heterogeneous nuclear ribonucleoprotein U-like 1
Gene Aliases
E130317O14Rik, E1B-AP5, E1BAP5, Hnrnpul, Hnrpul1
Chromosome Location
Chr.7: 25721165 - 25756245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144922.2 NP_659171.1 5 - 6 874 71
NM_178089.2 NP_835190.1 5 - 6 855 71
GenBank mRNA AK042090.1 - 5 - 6 855 71
AK087545.1 - 5 - 6 844 71
AK143666.1 - 5 - 6 917 71
AK155604.1 - 5 - 6 885 71
BC021506.1 - 5 - 6 898 71
BC027844.1 - 2 - 3 167 71
BC078642.1 - 5 - 6 798 71

View Full Product Details