Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf1
Entrez Gene ID
230584
Gene Name
Yip1 domain family, member 1
Gene Aliases
C030002N13Rik
Chromosome Location
Chr.4: 107314254 - 107359823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205156.1 NP_001192085.1 5 - 6 555 66
NM_145550.3 NP_663525.1 6 - 7 636 66
GenBank mRNA AK081205.1 - 6 - 7 623 66
AK157536.1 - 6 - 7 623 66
AK212572.1 - 1 - 2 102 66
AK214559.1 - 1 - 2 105 66
BC009080.1 - 6 - 7 568 66

View Full Product Details