The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145571.2 | NP_663546.1 | 4 - 5 | 627 | 102 |
GenBank mRNA | AK028416.1 | - | 4 - 5 | 650 | 102 |
AK158228.1 | - | 4 - 5 | 610 | 102 | |
AK167515.1 | - | 4 - 5 | 622 | 102 | |
AK170691.1 | - | 4 - 5 | 622 | 103 | |
AK172205.1 | - | 4 - 5 | 630 | 102 | |
AK216221.1 | - | 3 - 4 | 318 | 102 | |
BC009149.1 | - | 4 - 5 | 627 | 102 | |
BC033463.1 | - | 4 - 5 | 581 | 102 |