Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pphln1
Entrez Gene ID
223828
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
Chromosome Location
Chr.15: 93398350 - 93491913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146062.3 NP_666174.2 5 - 6 687 72
NM_175363.4 NP_780572.1 4 - 5 480 72
GenBank mRNA AK081606.1 - 4 - 5 473 72
AK150904.1 - 4 - 5 431 72
AK160138.1 - 4 - 5 453 72
BC031486.1 - 5 - 6 640 72

View Full Product Details