Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pphln1
Entrez Gene ID
223828
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
Chromosome Location
Chr.15: 93398350 - 93491913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083114.1 NP_001076583.1 7 - 8 819 63
NM_001285863.1 NP_001272792.1 7 - 8 948 63
NM_001285864.1 NP_001272793.1 6 - 7 727 63
NM_146062.3 NP_666174.2 9 - 10 1083 63
NM_175363.4 NP_780572.1 8 - 9 876 63
GenBank mRNA AK081606.1 - 8 - 9 869 63
AK150904.1 - 8 - 9 827 63
AK158488.1 - 7 - 8 949 63
AK160138.1 - 8 - 9 849 63
AK168443.1 - 7 - 8 821 63
AK184352.1 - 1 - 2 42 63
BC031486.1 - 9 - 10 1036 63
BC150854.2 - 6 - 7 723 63

View Full Product Details