Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nrd1
Entrez Gene ID
230598
Gene Name
nardilysin, N-arginine dibasic convertase, NRD convertase 1
Gene Aliases
2600011I06Rik, AI875733, NRD-C
Chromosome Location
Chr.4: 109000615 - 109061771 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146150.2 NP_666262.2 29 - 30 3229 81
GenBank mRNA AK031548.1 - 29 - 30 3229 81
AK050944.1 - 14 - 15 1328 81
AK166470.1 - 27 - 28 3228 81
BC023786.1 - 28 - 29 2607 81
BC024544.1 - 13 - 14 1214 81
BC026832.1 - 29 - 30 3279 81
BC029977.1 - 20 - 21 1865 81
BC036128.1 - 29 - 30 3347 81

View Full Product Details