Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdm1
Entrez Gene ID
17245
Gene Name
transformed mouse 3T3 cell double minute 1
Gene Aliases
Arrd2, Mdm-1
Chromosome Location
Chr.10: 118141537 - 118168999 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162904.1 NP_001156376.1 11 - 12 1789 71
NM_001162905.1 NP_001156377.1 10 - 11 1654 71
NM_010785.2 NP_034915.2 10 - 11 1759 71
NM_148922.3 NP_683724.2 10 - 11 1759 71
GenBank mRNA AK004789.2 - 10 - 11 1714 71
AK011769.1 - 10 - 11 1714 71
BC052680.1 - 10 - 11 1709 71
BC137608.1 - 11 - 12 1717 71
BC144777.1 - 11 - 12 1717 71
EU490427.1 - 11 - 12 1631 71
EU490428.1 - 10 - 11 1496 71
M20823.1 - 10 - 11 1622 71

View Full Product Details