The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007414.3 | NP_031440.1 | 2 - 3 | 517 | 65 |
GenBank mRNA | AF244347.1 | - | - | 486 | 65 |
AK010501.1 | - | 2 - 3 | 483 | 65 | |
AK146280.1 | - | 2 - 3 | 487 | 65 | |
AK153498.1 | - | 2 - 3 | 482 | 65 | |
AK171798.1 | - | 2 - 3 | 487 | 65 | |
BC003437.1 | - | 2 - 3 | 475 | 65 | |
BC058232.1 | - | 2 - 3 | 474 | 65 | |
L13290.1 | - | 1 - 2 | 314 | 65 |