Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ofcc1
Entrez Gene ID
218165
Gene Name
orofacial cleft 1 candidate 1
Gene Aliases
opo
Chromosome Location
Chr.13: 40001834 - 40361259 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF548110.1 - 1 - 2 269 63

View Full Product Details