The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172409.2 | NP_765997.2 | 24 - 25 | 3316 | 67 |
GenBank mRNA | AF513716.1 | - | 13 - 14 | 1713 | 67 |
AK080318.1 | - | 4 - 5 | 491 | 67 | |
AK173293.1 | - | 7 - 8 | 663 | 67 | |
BC048004.1 | - | 9 - 10 | 1204 | 67 | |
BC064731.1 | - | 4 - 5 | 473 | 67 |