The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172410.2 | NP_765998.1 | 6 - 7 | 586 | 106 |
GenBank mRNA | AK010450.1 | - | 5 - 6 | 537 | 106 |
AK028864.1 | - | 6 - 7 | 587 | 106 | |
AK045380.1 | - | - | 445 | 106 | |
AK129055.1 | - | 5 - 6 | 536 | 106 | |
AK167105.1 | - | 5 - 6 | 537 | 106 | |
AK169194.1 | - | 3 - 4 | 307 | 106 | |
AK169693.1 | - | 5 - 6 | 556 | 106 | |
BC023140.1 | - | 5 - 6 | 532 | 106 |