Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clk3
Entrez Gene ID
102414
Gene Name
CDC-like kinase 3
Gene Aliases
AI256811
Chromosome Location
Chr.9: 57750621 - 57765860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007713.4 NP_031739.3 2 - 3 942 109
GenBank mRNA AF033565.1 - 2 - 3 218 109
AK090215.1 - 2 - 3 183 109
AK133936.1 - 2 - 3 188 109
AK136481.1 - 2 - 3 205 109
AK140015.1 - 2 - 3 185 109
AK145880.1 - 2 - 3 197 109
AK146942.1 - 2 - 3 945 109
AK154003.1 - 2 - 3 191 109
AK167300.1 - 2 - 3 944 109
BC085084.1 - 2 - 3 185 109

View Full Product Details