Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gorasp2
Entrez Gene ID
70231
Gene Name
golgi reassembly stacking protein 2
Gene Aliases
0610011A07Rik, 2410043M02Rik, 5730520M13Rik, 9430094F20Rik, AW552058, GOLPH2, GRASP55, GRS2, p59
Chromosome Location
Chr.2: 70661489 - 70691725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027352.4 NP_081628.3 2 - 3 326 83
NR_027343.1 3 - 4 523 83
GenBank mRNA AK002515.1 - 3 - 4 405 83
AK010664.2 - 2 - 3 246 83
AK033413.1 - 2 - 3 260 83
AK044691.1 - - 211 83
AK077153.1 - 2 - 3 202 83
AK077683.1 - 2 - 3 261 83
AK077706.1 - 2 - 3 193 83
AK088239.1 - 2 - 3 208 83
AK148337.1 - 2 - 3 260 83
AK151843.1 - 2 - 3 208 83
AK152475.1 - 2 - 3 208 83
AK152544.1 - 2 - 3 208 83
AK152603.1 - 2 - 3 236 83
AK153049.1 - 2 - 3 208 83
AK153555.1 - 2 - 3 260 83
AK159469.1 - 2 - 3 258 83
AK167823.1 - 2 - 3 260 83
AK168928.1 - 3 - 4 456 83
AK169382.1 - 2 - 3 208 83
AK210658.1 - 1 - 2 84 83
AK211119.1 - 1 - 2 84 83
AK216284.1 - 1 - 2 84 83
AK219584.1 - 1 - 2 84 83
BC005600.1 - 2 - 3 199 83
BC016455.1 - 1 - 2 51 83
BC024982.1 - 2 - 3 212 83
BU707592.1 - 2 - 3 326 83

View Full Product Details