Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgm2l1
Entrez Gene ID
70974
Gene Name
phosphoglucomutase 2-like 1
Gene Aliases
4931406N15Rik, AI553438, BM32A
Chromosome Location
Chr.7: 100227283 - 100278872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027629.3 NP_081905.1 4 - 5 789 95
GenBank mRNA AK016437.1 - 2 - 3 148 95
AK039189.1 - 4 - 5 773 95
AK040513.1 - 4 - 5 709 95
BC020351.1 - 4 - 5 1002 95
BC076571.1 - 4 - 5 686 95

View Full Product Details