Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uaca
Entrez Gene ID
72565
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
2700059D02Rik, nucling
Chromosome Location
Chr.9: 60793206 - 60880370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028283.2 NP_082559.1 10 - 11 967 96
GenBank mRNA AB030647.1 - 10 - 11 1037 96
AK148900.1 - 10 - 11 1073 96
BC033470.1 - 10 - 11 884 96
BC042415.1 - 10 - 11 967 96

View Full Product Details