The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172261.3 | NP_758465.2 | 6 - 7 | 2357 | 61 |
GenBank mRNA | AY508450.1 | - | 6 - 7 | 2357 | 61 |
BC029672.1 | - | 5 - 6 | 1128 | 61 |