Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc14b
Entrez Gene ID
218294
Gene Name
CDC14 cell division cycle 14B
Gene Aliases
2810432N10Rik, A530086E13Rik, AA472821, CDC14B3, Cdc14B1
Chromosome Location
Chr.13: 64189271 - 64275290 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122989.1 NP_001116461.1 11 - 12 1211 91
NM_172587.3 NP_766175.3 11 - 12 1475 91
GenBank mRNA AK041155.1 - 11 - 12 1197 91
AK044332.1 - 11 - 12 1777 91
AK161345.1 - 11 - 12 1476 91
BC049794.1 - 11 - 12 1417 91
BC057357.1 - 11 - 12 1366 91

View Full Product Details