The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027060.1 | NP_081336.1 | 9 - 10 | 1738 | 64 |
NM_172618.2 | NP_766206.1 | 9 - 10 | 1738 | 64 | |
GenBank mRNA | AK006275.1 | - | 4 - 5 | 624 | 64 |
AK029578.1 | - | 9 - 10 | 1901 | 64 | |
AK029907.1 | - | 2 - 3 | 378 | 64 | |
AK052664.1 | - | 9 - 10 | 1622 | 64 | |
AK153973.1 | - | 9 - 10 | 1665 | 64 | |
BC037031.1 | - | 7 - 8 | 1205 | 64 | |
BC057897.1 | - | 9 - 10 | 1616 | 64 |