Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbln2
Entrez Gene ID
12405
Gene Name
cerebellin 2 precursor protein
Gene Aliases
6330593N19Rik, A730004O05
Chromosome Location
Chr.18: 86711053 - 86718285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK032112.1 - 1 - 2 384 70
AK042555.1 - 2 - 3 659 70
BC055682.1 - 3 - 4 657 70

View Full Product Details