The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172653.2 | NP_766241.2 | 7 - 8 | 2125 | 56 |
GenBank mRNA | AK030685.1 | - | - | 261 | 56 |
AK049099.1 | - | 5 - 6 | 871 | 56 | |
AK122483.1 | - | 6 - 7 | 1559 | 56 | |
AK136117.1 | - | 6 - 7 | 1837 | 56 | |
BC052880.1 | - | 7 - 8 | 2239 | 56 | |
BC059214.1 | - | 5 - 6 | 1009 | 56 | |
BC062918.1 | - | 7 - 8 | 2125 | 56 |