The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172671.2 | NP_766259.2 | 17 - 18 | 2051 | 98 |
GenBank mRNA | AK033699.1 | - | 2 - 3 | 171 | 98 |
AK044357.1 | - | 16 - 17 | 1359 | 98 | |
BC056637.1 | - | 2 - 3 | 188 | 98 |