The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172694.2 | NP_766282.1 | 1 - 2 | 707 | 67 |
GenBank mRNA | AK035987.1 | - | 1 - 2 | 708 | 67 |
AK039319.1 | - | 1 - 2 | 708 | 67 | |
AK122378.1 | - | - | 698 | 67 | |
BC088994.1 | - | 1 - 2 | 616 | 67 | |
BC138128.1 | - | 1 - 2 | 646 | 67 | |
BC138129.1 | - | 1 - 2 | 646 | 67 |