The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290696.1 | NP_001277625.1 | 7 - 8 | 1034 | 88 |
NM_172699.3 | NP_766287.1 | 8 - 9 | 1136 | 88 | |
GenBank mRNA | AK082840.1 | - | 8 - 9 | 1137 | 88 |
AK129271.1 | - | 7 - 8 | 1007 | 88 | |
AK139508.1 | - | 8 - 9 | 1070 | 88 | |
BC058231.1 | - | 8 - 9 | 1109 | 88 |