The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172716.4 | NP_766304.2 | 6 - 7 | 573 | 88 |
GenBank mRNA | AK033609.1 | - | 6 - 7 | 648 | 88 |
AK085574.1 | - | 6 - 7 | 629 | 88 | |
AK089091.1 | - | 6 - 7 | 574 | 88 | |
BC049266.1 | - | - | 440 | 88 |