Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr3
Entrez Gene ID
234549
Gene Name
HEAT repeat containing 3
Gene Aliases
C030036P15Rik
Chromosome Location
Chr.8: 88137885 - 88171952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172757.3 NP_766345.3 5 - 6 753 72
GenBank mRNA AK047782.1 - 5 - 6 784 72
AK052635.1 - 5 - 6 780 72
AK134510.1 - 5 - 6 758 72
AK136424.1 - 5 - 6 763 72
AK140522.1 - 5 - 6 771 72
AK163715.1 - 5 - 6 740 72
AK165541.1 - 5 - 6 647 72
BC030373.1 - 1 - 2 98 72
BC065060.1 - 5 - 6 753 72

View Full Product Details