Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr3
Entrez Gene ID
234549
Gene Name
HEAT repeat containing 3
Gene Aliases
C030036P15Rik
Chromosome Location
Chr.8: 88137885 - 88171952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172757.3 NP_766345.3 14 - 15 2057 85
GenBank mRNA AK047782.1 - 14 - 15 2088 85
AK052635.1 - 14 - 15 2084 85
AK134510.1 - 14 - 15 2062 85
AK136424.1 - 14 - 15 2067 85
AK140522.1 - 14 - 15 2075 85
AK163533.1 - 1 - 2 133 85
AK163715.1 - 14 - 15 2820 85
AK165541.1 - 14 - 15 1951 85
BC030373.1 - 10 - 11 1402 85
BC048074.1 - 3 - 4 775 85
BC065060.1 - 14 - 15 2057 85

View Full Product Details